Sodium dodecyl sulfate (SDS) FCC
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
Sodium Lauryl Sulfate, FCC is a surfactant and emulsifier commonly used in food products. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
GHS hazard : Skull and crossbones;Exclamation mark
UN2811
Formula:
CH₃(CH₂)₁₁OSO₃Na Storage Temperature: Ambient |
MDL Number:
MFCD00036175 CAS Number: 151-21-3 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Assay | 59.0 % min. |
Alkalinity | To Pass Test(0.2 |
Sodium Chloride and Sodium Sulfate | max. 8.0 % |
Lead (Pb) | max. 2 mg/kg |
Unsulfated Alcohols | max. 4.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...