Sodium hydroxide ≥95.0%, pellets FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

S1303-12KG S1303-2.5KG S1303-2.5KG-CS S1303-500GM S1303-500GM-CS S1303-50KGBL
75812-234EA 939.75 USD
75812-234 75812-236 75812-238 75812-240 75812-242 75812-244
Sodium hydroxide ≥95.0%, pellets FCC
Sodium hydroxide

Sodium Hydroxide, Pellets, FCC is used in the chemical washing and peeling of fruits and vegetables and as a thickening agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Corrosion;Exclamation mark
UN1823

Formula: NaOH
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8,II

Order Now

Specification Test Results

Assay (Total Alkali As NaOH) 95.0 - 100.5 %
Arsenic (As) max. 3 mg/kg
Carbonate (As Na₂CO₃) max. 3.0 %
Insoluble Substances & Organic Matter To Pass Test
Lead (Pb) max. 2 mg/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR