Sodium hydroxide ≥95.0%, pellets FCC
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
Sodium Hydroxide, Pellets, FCC is used in the chemical washing and peeling of fruits and vegetables and as a thickening agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
GHS hazard : Corrosion;Exclamation mark
UN1823
Formula:
NaOH Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 UN: 1823 ADR: 8,II |
Specification Test Results
Assay (Total Alkali As NaOH) | 95.0 - 100.5 % |
Arsenic (As) | max. 3 mg/kg |
Carbonate (As Na₂CO₃) | max. 3.0 % |
Insoluble Substances & Organic Matter | To Pass Test |
Lead (Pb) | max. 2 mg/kg |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...