Saccharin ≥98.0%, powder FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: o-Sulphobenzimide, o-Benzoic sulphimide, o-Benzoic sulfimide, o-Sulfobenzimide

S1003-500GM S1003-125GM S1003-2.5KG
75811-894EA 567.33 USD
75811-894 75812-118 75812-120
Saccharin ≥98.0%, powder FCC
Saccharin

Saccharin, Powder, FCC is used as an artificial sweetener. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: C₇H₅NO₃S
MW: 183.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005866
CAS Number: 81-07-2

Order Now

Specification Test Results

Assay (C₇H₅NO₃S; Dried Basis) 98.0 - 101.0 %
Melting Range 226° - 230°C
Benzoic and Salicylic Acids To Pass Test
Lead max. 2 mg/kg
Loss On Drying max. 1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR