Poloxamer 407 (Pluronic® F-127) NF
Supplier: Spectrum Chemical Mfg. Corp.
Poloxamer 407, NF is a national formulary approved surfactant that finds utility because of its amphiphilic structure.
- Amphphilic structure
- Increases miscibility
- Controls thickness of personal care products
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
Formula:
HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)aH Storage Temperature: Ambient |
MDL Number:
MFCD00082049 CAS Number: 9003-11-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...