Poloxamer 407 (Pluronic® F-127) NF

Supplier: Spectrum Chemical Mfg. Corp.

P1166-45KGBL P1166-2.5KG P1166-500GM P1166-12KG P1166-25KGBL
700011-996EA 4839.07 USD
700011-996 95034-970 95034-972 75811-800 75811-802
Poloxamer 407 (Pluronic® F-127) NF
Poloxamer 407 (Pluronic® F-127)

Poloxamer 407, NF is a national formulary approved surfactant that finds utility because of its amphiphilic structure.


  • Amphphilic structure
  • Increases miscibility
  • Controls thickness of personal care products


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)aH
Storage Temperature: Ambient
MDL Number: MFCD00082049
CAS Number: 9003-11-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR