DL-Malic acid ≥99.0% FCC

Supplier: Spectrum Chemical Mfg. Corp.

MA147-500GM MA147-2.5KG MA147-12KG MA147-45KGBL
95032-976EA 146.03 USD
95032-976 95032-974 75811-704 75811-706
DL-Malic acid ≥99.0% FCC
DL-Malic acid
Formula: HOOCCH(OH)CH₂COOH
Storage Temperature: Ambient
MDL Number: MFCD00064212
CAS Number: 617-48-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=920b54632a869bc5df22624fb9fecd4d11cae65e&vid=278193678126274041defdc0bd71c7c40efb3085&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR