Magnesium stearate BP, JP, NF

Supplier: Spectrum Chemical Mfg. Corp.

MA130-12KG MA130-500CS6 MA130-2.5CS4 MA130-2.5KG MA130-500GM MA130-45KGBL
700005-416EA 1962.53 USD
700005-416 95034-260 95034-256 95034-258 95034-262 75811-702
Magnesium stearate BP, JP, NF
Magnesium stearate

Magnesium Stearate, NF, BP, JP is used as an anti-adherent in the manufacture of of medical capsules, tablets, and powders.


All Spectrum Chemical NF, BP, JP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: C₃₆H₇₀MgO₄
Storage Temperature: Ambient
MDL Number: MFCD00036391
CAS Number: 557-04-0

Order Now

Specification Test Results

Assay (As MG; Dried Basis) 4.0 - 5.0%
Relative Content of Stearic & Palmitic Acid To Pass Test
Total Aerobic Microbial Count max. 1000 Cfu/G
Total Combined Molds & Yeasts Count max. 500 Cfu/G


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR