Magnesium sulfate heptahydrate ≥99.5% FCC
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
sulphuric acid magnesium salt heptahydrate, sulfuric acid magnesium salt heptahydrate, Epsom salt, Bitter salt
Magnesium Sulfate, Heptahydrate, FCC, commonly called Epsom salt, is used as a brewing salt in beer production and a tofu coagulant. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
MgSO₄·7H₂O MW: 246.48 g/mol Melting Pt: 1124 Storage Temperature: Ambient |
MDL Number:
MFCD00149785 CAS Number: 10034-99-8 Merck Index: 13,05715 |
Specification Test Results
Assay (After Ignition) | 99.5 % min. |
Loss On Ignition | 40.0 - 52.0 % |
Lead (Pb) | max. 4 mg/kg |
Selenium (Se) | max. 0.003 % |
Identification | To Pass Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...