D-Lactose monohydrate, spray-dried powder NF

Supplier: Spectrum Chemical Mfg. Corp.

LA105-12KG LA105-500GM LA105-2.5KG LA105-25KGBL
700000-246EA 911.75 USD
700000-246 95033-806 95033-804 75811-526
D-Lactose monohydrate, spray-dried powder NF
D-Lactose monohydrate

Lactose, Monohydrate, Spray-Dried Powder, NF is used to help form tablets because it has excellent compressibility properties and as a diluent powder for dry-powder inhalations. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: C₁₂H₂₂O₁₁·1H₂O
Storage Temperature: Ambient
MDL Number: MFCD00150747
CAS Number: 64044-51-5

Order Now

Specification Test Results

Clarity and Color of Solution To Pass Test
Specific Rotation [A]D +54.4 +55.9
Acidity or Alkalinity To Pass Test
Water Determination 4.5 - 5.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR