Glycine ≥98.5% FCC
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Aminoacetic acid, Glycocoll, H-Gly-OH
Glycine, FCC is an organic compound and is also the smallest of the 20 common amino acids in proteins. It is a sweet tasting, colorless, crystalline solid. Glycine is a common building block to many natural products and is a precursor to proteins It is also used as an additive in animal food, and as a food supplement and sweetener in food destined for human consumption. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
NH₂CH₂COOH Storage Temperature: Ambient |
MDL Number:
MFCD00008131 CAS Number: 56-40-6 |
|
Specification Test Results
Assay (C₂H₅No₂); Dried Basis) | 98.5 - 101.5 % |
Lead (Pb) | max.5 mg/kg |
Loss On Drying | max. 0.2 % |
Residue On Ignition | max. 0.1 % |
Insoluble Foreign Matter in Amino Acids | To Pass Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...