Eucalyptus oil FCC

Supplier: Spectrum Chemical Mfg. Corp.

EU106-100ML EU106-200LTBL EU106-20LTBL EU106-4LTGL EU106-4LTGL-CS EU106-500MLGL
75811-096EA 79.85 USD
75811-096 75811-098 75811-100 75811-102 75811-104 75811-106
Eucalyptus oil FCC
Eucalyptus oil

Eucalyptus Oil, FCC is a fragarance and flavor additive. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Flame;Health hazard;Exclamation mark UN1197

Storage Temperature: Ambient
MDL Number: MFCD00146880
CAS Number: 8000-48-4
UN: 1197
ADR: 3,III

Order Now

Specification Test Results

Assay (As Eucalyptol: C₁₀H₁₈O) 70.0 % min.
Solubility in 70% Alcohol (V/V) To Pass Test
Specific Gravity @ 25°C 0.905- 0.925
Refractive Index @ 20°C 1.458- 1.470
Phellandrene To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR