Alcohol ≥94.9-96% USP (190 Proof)

Supplier: Spectrum Chemical Mfg. Corp.

ET108-4LTGL ET108-4LTGL-CS
700000-282EA 773.24 USD
700000-282 75811-092
Alcohol ≥94.9-96% USP (190 Proof)
Alcohol

Alcohol, 190 Proof, USP is approximately 95% Ethanol and 5% water. This product, 190 proof USP grade alcohol, is used as an antiseptic to disinfect skin prior to injections and as a solvent. All Spectrum Chemical USP products, including all USP alcohol products, are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


GHS hazard : Flame;Health hazard;Exclamation mark
UN1170

Formula: H₃CCH₂OH
Density: 0.816
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3,II

Order Now

Specification Test Results

Assay (By Volume) 94.9 - 96.0 %
Clarity of Solution To Pass Test
Color of Solution To Pass Test
Specific Gravity @ 15.56°C 0.812- 0.816
Acidity or Alkalinity To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR