Corn oil NF

Supplier: Spectrum Chemical Mfg. Corp.

CO136-500MLG CO136-200LTB CO136-4LTGL CO136-25ML
700000-132EA 1021.83 USD
700000-132 700000-136 700000-130 700000-134 75810-946
Corn oil NF
Corn oil

Corn Oil, NF its uses include as a solvent vehicle for topical preparations and as a glidant excipient for oral dosage tablets and capsules. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Storage Temperature: Ambient
MDL Number: MFCD00130865
CAS Number: 8001-30-7

Order Now

Specification Test Results

Fatty Acid Composition To Pass Test
Acid Value max. 0.2
Peroxide Value max. 10.0
Water Determination max. 0.1 %
Alkaline Impurities To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR