Citric acid, anhydrous ≥99.5%, granular USP
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid
CI133-45KGBL
CI133-2.5KG
CI133-12KG
CI133-500GM
CI133-25KGBL
89078-740EA
1635.15
USD
89078-740
95032-586
89078-738
95032-588
75810-940
Citric acid, anhydrous ≥99.5%, granular USP
Citric acid
Anhydrous Citric Acid, Granular, USP belongs to a class of drugs known as urinary alkalinizers that are used to treat certain metabolic problems (acidosis) caused by kidney disease. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
GHS hazard : Exclamation mark
Formula:
HOC(COOH)(CH₂COOH)₂ MW: 192.13 g/mol Boiling Pt: 310 Storage Temperature: Ambient |
MDL Number:
MFCD00011669 CAS Number: 77-92-9 Merck Index: 13,02350 |
|
Specification Test Results
Assay (Anhydrous Basis) | 99.5 - 100.5 % |
Clarity of Solution | To Pass Test |
Color of Solution | To Pass Test |
Water | max. 1.0 % |
Residue On Ignition | max. 0.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...