Calcium citrate tetrahydrate ≥97.5%, Powder FCC
Supplier: Spectrum Chemical Mfg. Corp.
Calcium Citrate, Tetrahydrate, Powder, FCC is an odorless white powder and the calcium salt of citric acid and is commonly used as a preservative and flavorant food additive. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₁₂H₁₀Ca₃O₁₄·4H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00150786 CAS Number: 5785-44-4 |
Specification Test Results
Assay [Ca₃(C₆H₅O₇)₂] | 97.5 - 100.5 % |
Loss On Drying | 10.0 - 14.0 % |
Fluoride | max. 0.003 % |
Lead (Pb) | max. 2 mg/kg |
Identification | To Pass Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...