Calcium chloride dihydrate ≥99.0% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Calcium Chloride Dihydrate, Calcium dichloride

C1099-2.5KG C1099-500GM C1099-12KG C1099-50KGBL
95032-242EA 750.55 USD
95032-242 95032-244 75810-744 75810-746
Calcium chloride dihydrate ≥99.0% FCC
Calcium chloride dihydrate

Calcium Chloride, Dihydrate, FCC is used as a sequestrant and firming agent in food products. It is considered as generally recognized as safe (GRAS) by the U.S. Food and Drug Administration. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


GHS hazard : Exclamation mark

Formula: CaCl₂·2H₂O
MW: 147.01 g/mol
Boiling Pt: 1600
Melting Pt: 176
Storage Temperature: Ambient
MDL Number: MFCD00149613
CAS Number: 10035-04-8
Merck Index: 13,01660

Order Now

Specification Test Results

Assay 99.0 - 107.0 %
Arsenic (As) max. 3 mg/kg
Fluoride max. 0.004 %
Lead (Pb) max. 5 mg/kg
Magnesium and Alkali Salts max. 4.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR