Benzoic acid ≥99.5% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

B1102-500GM B1102-2.5KG B1102-12KG B1102-45KGBL
95031-988EA 229.35 USD
95031-988 95031-986 75810-666 75810-668
Benzoic acid ≥99.5% FCC
Benzoic acid

Benzoic Acid, FCC is used as a food preservative inhibiting the growth of mold, yeast and some bacteria. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


GHS hazard : Exclamation mark

Formula: C₇H₆O₂
Storage Temperature: Ambient
MDL Number: MFCD00002398
CAS Number: 65-85-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d561f311b11ea8ab2c8f07253e6748e588eb71aa&vid=dcf5e45072cda5106216b497cd29716270ae772c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (C₇H₆O₂) 99.5 - 100.5 %
Solidification Point 121° - 123°C
Lead max. 2.0 mg/kg
Readily Carbonizable Substances To Pass Test
Readily Oxidizable Substances To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR