Acetone ≥99.0% NF

Supplier: Spectrum Chemical Mfg. Corp.

AC115-200LTB AC115-1LTGL AC115-20LTMT AC115-4LTGL AC115-500MLG AC115-100ML AC115-4LTPL AC115-1LTGL-CS AC115-4LTGL-CS AC115-4LTPL-CS AC115-500ML-CS
700003-862EA 1749.25 USD
700003-862 700003-860 700003-864 700003-866 700003-870 700003-858 700003-868 75810-598 75810-600 75810-602 75810-604
Acetone ≥99.0% NF
Acetone

Acetone, NF is a low boiling, volatile organic compound. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: CH₃COCH₃
Density: 0.788
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
UN: 1090
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay [(CH₃)₂CO] 99.0 % min.
Specific Gravity @ 25°C max. 0.789
Water max. 0.5 %
Nonvolatile Residue max. 0.004 %
Readily Oxidizable Substances To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR