L(+)-Arginine ≥98.5% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: H-L-Arg-OH, (S)-2-Amino-5-guanidinopentanoic acid, Arginine, (S)-(+)-Arginine, L-Arginine, H-Arg-OH

A1336-100GM A1336-1KG A1336-50KGBL
95033-846EA 205.91 USD
95033-846 95033-848 75810-524
L(+)-Arginine ≥98.5% FCC
L(+)-Arginine

L-Arginine, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₆H₁₄N₄O₂
MW: 174.2 g/mol
Boiling Pt: 368
Melting Pt: 238
Storage Temperature: Ambient
MDL Number: MFCD00002635
CAS Number: 74-79-3
Merck Index: 13,00785

Order Now

Specification Test Results

Assay (Dried Basis) 98.5 - 101.5 %
Specific Rotation @ 20°C +26.0 to +27.9
Lead (Pb) max. 5 mg/kg
Loss On Drying max. 1.0 %
Residue On Ignition max. 0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR