Aluminium sulfate octadecahydrate ≥99.5%, crystals FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Trade termforalum, Aluminum sulfate octadecahydrate, Aluminum(III) sulfate octadecahydrate, Aluminum(III) sulphate octadecahydrate, Aluminum sulphate octadecahydrate

A1114-45KGBL
75810-450EA 1390.65 USD
75810-450
Aluminium sulfate octadecahydrate ≥99.5%, crystals FCC
Aluminium sulphate octadecahydrate

Aluminum Sulfate, Crystal, FCC is soluble in water and is mainly used as a thickener and emulsifier in food preparation. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark UN3077

Formula: Al₂(SO₄)₃·18H₂O
MW: 666.43 g/mol
Melting Pt: 86
Storage Temperature: Ambient
MDL Number: MFCD00149136
CAS Number: 7784-31-8
UN: 3077
ADR: 9 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4dace68759944aa1c6c0f56a372ea396f004e261&vid=e63de3c58650a97622a605c237aa52402ff157e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay [Al₂(SO₄)₃] 99.5-114.0 %
Alkalies and Alkaline Earths To Pass Test(Lim
Ammonium Salts To Pass Test
Fluoride max. 0.003 %
Lead (Pb) max. 3 mg/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR