Paraffin wax blended with synthetic polymers, Solidification point: 56°C, Surgipath® Paraplast Plus
Supplier: Leica Biosystems
39602004
75810-408CS 199.67 USD
75810-408
Paraffin wax blended with synthetic polymers, Solidification point: 56°C, Surgipath® Paraplast Plus
Paraffin wax blended with synthetic polymers
Paraplast Plus is suggested for large tissue or tissue that may normally present infiltration issues. Paraplast Plus contains a small percentage of dimethyl sulfoxide (DMSO), allowing accelerated tissue penetration. It has a melting point of 56° C.
CAS Number: 8002-74-2