Hematoxylin, powder, Surgipath®, certified by the Biological Stain Commission

Supplier: Leica Biosystems

3803815
75810-366EA 228.57 USD
75810-366
Hematoxylin, powder, Surgipath®, certified by the Biological Stain Commission
Hematoxylin

Powdered Stain Hematoxylin Dye can be used to prepare all Hematoxylin formulations for routine and special staining procedures that require nuclear staining.


  • Certified by the Biological Stain Commission
  • Colour Index Number 75290


The total dye content is stated on each label.


Certifications: Dye is certified by the Biological Stain Commission.

Formula: C₁₆H₁₄O₆
MDL Number: MFCD00078111
CAS Number: 517-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR