Orange G solution, Surgipath® for cytology

Supplier: Leica Biosystems
Danger

3801660
75810-356EA 190.2 USD
75810-356
Orange G solution, Surgipath® for cytology
Orange G

Orange G-6 is an alcohol-based stain used in the cytology laboratory to detect the presence of keratin in the cytoplasm.


  • Alcohol-based
  • Detects keratin in cytoplasm


It is manufactured by Leica Biosystems under the strictest controls using a standard formulation for conventional GYN/Non-GYN staining applications.

Formula: C₁₆H₁₀N₂Na₂O₇S₂
MDL Number: MFCD00012457
CAS Number: 1936-15-8
UN: 1987
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR