2-Propanol, anhydrous, Surgipath® for histology and pathology

Supplier: Leica Biosystems
Danger

3803660
75810-350CS 179.07 USD
75810-350
2-Propanol, anhydrous, Surgipath® for histology and pathology
2-Propanol

Isopropanol ispure filtered and laboratory tested


  • High-quality filtered Isopropanol 99% Anhydrous
  • ISO certified
  • Available in non‑breakable, gallon containers


The Isopropanol is recommended for small tissue and biopsy processing as well as any other laboratory procedures requiring Isopropanol, including Xylene-free processing aplication.


Certifications: ISO certified

Formula: (CH₃)₂CHOH
Boiling Pt: 82 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR