Anti-MMP3 Mouse Monoclonal Antibody [clone: 1F5A9]

Supplier: Proteintech
66338-1-IG-150UL
75806-496EA 462.47 USD
75806-496
Anti-MMP3 Mouse Monoclonal Antibody [clone: 1F5A9]
Antibodies
Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis.

  • Positive WB detected in COLO 320 cells, fetal human brain tissue, pig heart tissue
  • Positive IHC detected in human breast cancer tissue, human stomach cancer tissue
  • Dilutions of WB : 1:1000 - 1:10000 and IHC : 1:50 - 1:500

This gene encodes an enzyme which degrades fibronectin, laminin, collagens III, IV, IX, and X, and cartilage proteoglycans. The enzyme is thought to be involved in wound repair, progression of atherosclerosis, and tumor initiation.

Type:
Antigen: MMP3 (matrix metallopeptidase 3 (stromelysin 1, progelatinase))
Clonality: Monoclonal
Clone: 1F5A9
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR