Amyloid Stain Kit, Congo Red, Polysciences
Supplier: POLYSCIENCES INC-RESEARCH CHEM
Amyloid stain kit used in the detection of amyloid in formalin fixed paraffin embedded (FFPE) tissue sections as well as frozen tissue.
- Recommended section thickness is 10 microns
Delivery information: Kit contains 250 mL bottle of each, congo red solution A, alkaline alcohol solution B, Gill's hematoxylin #3 Solution C.