eLITE Xenon Set, UVP
Supplier: Analytik Jena US
98-0105-01
75803-210EA 12434.51 USD
75803-210
eLITE Xenon Set, UVP
Lights Spectrum Lamps
For use with Analytik Jena US Advanced Imaging Systems.
The UVP eLITE Light Source uses Xenon light engine technology to provide uniform and high-intensity light across the full range of brilliant mulispectral excitation which can be produced for a variety of fluorescent dyes.