arium® mini Scientific Kit Ultrapure Cartridge, Sartorius Corp.
Supplier: Sartorius
H2O-S-PACK
75803-116EA 570.34 USD
75803-116
arium® mini Scientific Kit Ultrapure Cartridge, Sartorius Corp.
Filters Liquid Process Filters
Deionization cartridge featuring top-down flow technology.
  • High performance capacity due to efficient ion exchange resins
  • Fast and effective adsorption of impurities by high-grade activated carbon
  • Optimized flow prevents separation of the mixed-bed resin
  • Patented connection method; easy exchange of consumables
The cartridge kits have been optimized for removal of both organic and inorganic constituents. Each kit has been designed specifically to match the particular laboratory water system and delivers ultrapure water that exceeds the ASTM Type 1 quality standard. This consistently high-quality water ensures optimal reproducibility of your results. Optimized filling materials, such as highly effective activated carbon along with exceptionally efficient ion exchange resins, ensure long-lasting performance and low-maintenance operation.The top-down technology provides ideal purification kinetics, preventing any mixing of cleaning media. The cartridge has been designed for enhanced flow rate in the cross section and optimal contact time with the medium.