arium® mini plus Pretreatment Cartridge, Sartorius Corp.
Supplier: Sartorius
H2O-CPR
75803-114EA 754.36 USD
75803-114
arium® mini plus Pretreatment Cartridge, Sartorius Corp.
Filters Liquid Process Filters
Reliable protection for the pretreatment of feed water.
  • Fast and effective adsorption of impurities by high-grade activated carbon
  • Highly efficient catalyzer for removing oxidating agents such as chlorine and ozone
  • Low-energy membranes for ecological and economic operation
Efficient purification is performed by a combination of activated carbon, a catalyzer and a downstream reverse osmosis membrane. The spherical, heavy metal ions and particulate contaminants, from feed water. In addition, due to the downstream reverse osmosis membrane, up to 98% of all salts, as well as bacteria and particles, are retained.