PYR Reagent, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z127
75803-062EA 47.17 USD
75803-062
PYR Reagent, Hardy Diagnostics
Microbiology Test Systems Enzymatic Tests
For use in conjunction with PYR disks for the detection of pyroglutamate aminopeptidase found in group A streptococci (Streptococcus pyogenes) and group D enterococci.
  • Combines with beta-naphthyl amide
  • Released during pyroglutamate aminopeptidase (PYR) activity to form a pink to red pigment
For use in conjunction with PYR disks for the detection of pyroglutamate aminopeptidase found in group A streptococci (Streptococcus pyogenes) and group D enterococci
The Hardy Diagnostics manufacturing facility and quality management system comply with the FDA's Good Manufacturing Practices (GMP) and have achieved ISO 13485 certification as a medical device manufacturer.
The PYR Reagent combines with beta-naphthyl amide released during pyroglutamate aminopeptidase (PYR) activity to form a pink to red pigment.