Hemin and Vitamin K Solution, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z237
75803-006PK 155.45 USD
75803-006
Hemin and Vitamin K Solution, Hardy Diagnostics
Media for Microbiology Additives and Supplements for Microbiology
A growth supplement for fastidious anaerobic bacteria.
(Hemin, 0.25mg/ml and Vitamin K, 0.050mg/ml) A growth supplement for fastidious anaerobic bacteria.
Ordering information: Minimum order of 5 packs