Hettich ROTOFIX 46H Heated Benchtop Centrifuge, Hettich Lab Technology
Supplier: HETTICH INSTRUMENTS
4600-51
75802-782EA 9026.18 USD
75802-782
Hettich ROTOFIX 46H Heated Benchtop Centrifuge, Hettich Lab Technology
Centrifuges Benchtop Centrifuges
Powerful and flexible benchtop centrifuge for organic and inorganic chemistry, preparatory and analytical chemistry, electroplanting, and petrochemistry
  • Industrial/environmental
  • Metal lid
  • Stainless steel centrifuging chamber
  • Viewing port in the lid
  • Twist lock for effortless opening and closing of the lid
  • Ergonomic control and information panel
  • 10 programmable memories
  • Easy rotor changing
  • Infinitely variable setting from +10 to +90°C (dependent on the ambient temperature)
  • Pre-heating function
Wide range of accessories with solutions for a variety of specialty applications. Universal swing-out rotor for tubes and bottles ranging from 3mL to 290mL. Quiet as a normal conversation. Will not increase noise levels or contribute to distractions. Easy to lift lid with front-facing control panel for effortless interaction and programming. Self-closing, auto-locking lid with steel latches. All-metal chamber provides reinforced security.
Certifications: Engineered and manufactured in accordance with all relevant safety and production standards.
Ordering information: Five-year manufacturer's warranty. For various available application packages, rotors, and accessories, please contact your Avantor representative.