Hettich ROTOFIX 46 Lab Benchtop Centrifuge, Hettich Lab Technology
Supplier: HETTICH INSTRUMENTS
4600-01
75802-780EA 8537.63 USD
75802-780
Hettich ROTOFIX 46 Lab Benchtop Centrifuge, Hettich Lab Technology
Centrifuges Benchtop Centrifuges
Hettich Lab Technology's ROTOFIX 46 is a powerful and flexible benchtop with standard and speciality applications including organic and inorganic chemistry, preparatory and analytical chemistry, electroplating and petrochemistry.
  • All-steel construction
  • General-purpose
  • Cytology
  • Quiet Operation – Quiet as a normal conversation
  • Will not increase noise levels or contribute to distractions
  • Functional Design –Easy to lift lid with front-facing control panel for effortless interaction and programming
  • Enhanced Safety –Self-closing, auto-locking lid with steel latches
  • All-metal chamber provides reinforced security
  • Five Year Warranty –All Hettich centrifuges are protected by our 5-year warranty and full technical support
  • Fully Certified – Engineered and manufactured in accordance with all relevant safety and production standards
  • Made in Germany – German engineered and manufactured quiet, reliable and safe since 1904
Certifications: Hettich Lab Technology centrifuges are manufactured under all relevant national and international safety standards and certifications.
Ordering information: All Hettich centrifuges include a 5-year warranty. Hettich Lab Technology centrifuge packages include everything you need to complete your application: centrifuge, rotor, buckets, adapters, and lids, where applicable. Supports over 210 application configurations; please contact your Avantor representative for more information.