Haematocrit Centrifuge, HAEMATOKRIT 200, Hettich Lab Technology
Supplier: HETTICH INSTRUMENTS
1801-01
75801-782EA 4041.06 USD
75801-782
Haematocrit Centrifuge, HAEMATOKRIT 200, Hettich Lab Technology
Centrifuges Benchtop Centrifuges
This compact centrifuge efficiently determines the erythrocyte volume fraction in blood
  • Metal lid and deep metal chamber which securely holds the rotor
  • Lid locking and holding; emergency lid lock release
  • Rotor lid also serves as an evaluation disc, enabling the haematocrit values to be read off directly after sedimentation
  • Practical control panel allows time and speed to be set quickly and easily
It can hold up to 24 capillaries. The maximum packing density of the cells is achieved within seven minutes, the cell-free plasma is then available for further tests. Suitable for use in neonatology, haematology and sports medicine.