Corning® Gosselin™ Round Petri Dish with 6 Vents, Corning
Supplier: Corning
BP53-03
75801-452CS 528.9 USD
75801-452
Corning® Gosselin™ Round Petri Dish with 6 Vents, Corning
Culture Dishes Petri Dishes
The Corning® Gosselin™ Round Petri Dish is manufactured from crystal polystyrene, and designed for use in microbiology testing applications
  • Crystal polystyrene construction
  • Double outerbag packaging
  • Easy opening sleeves
  • Sterilized to SAL 10¯6
  • Optimal flatness
The dish is designed with optimal flatness and features six vents. Manufactured in a class 100 environment.
Certifications: Food contact : Raw material is compliant with regulation EC N°1935/2004.
Sterility Assurance Level based on ISO 11137.