VWR® 25D Digital Homogenizer
Supplier: VWR International
75800-448
75800-448EA 4907.58 USD
75800-448
VWR® 25D Digital Homogenizer
Homogenizers Benchtop Homogenizers
The versatile and powerful VWR® 25D Homogenizer is ideal for a wide range of lab applications, including but not limited to compounding, emulsions and suspensions, cell disruption, protein extractions, medical research or pharmaceutical processing.
  • Compact digital homogenizer unit with high watt motor
  • 0.05ml - 3.5 L volume range and viscosity up to 50,000 mPas
  • Bright LED display and user-friendly controls
  • Can homogenize, emulsify, blend and/or mix
  • Includes U-shape post mounted stand assembly
  • VWR® Generator probe sold separately
The VWR® 25D Homogenizer has a wide volume capability with processing up to 3.5 Liters (higher sample volumes dependent on viscosity). Its digital display and speed control provides the ability to fine-tune speeds between 0 to 30,000 RPM, allowing its speed adjustment to be precisely controlled within the range.
Certifications: CE marked.
Accessories information: Generator probes must be purchased with homogenizer units.
Ordering information: Supplied with a two-year manufacturer's warranty. VWR® Generator probe(s) are required for use with the VWR25D Homogenizer model and are sold separately (VWR® GenPack Adapter is required for VWR® GenPack Generator Probes).