Urine Clean Catch Easy Grip Handle, Azer Scientific
Supplier: Azer Scientific Inc.
ES64310
75799-876CS 212.99 USD
75799-876
Urine Clean Catch Easy Grip Handle, Azer Scientific
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
Easy Grip Handle for 90 mL and 120 mL urine containers.
  • Compatible with both 90 mL and 120 mL urine containers
  • Provides patients with a clean, easy alternative during urine collection
Offer this handle during urine clean catch collection to prevent contamination. The handle simply slides up the bottom of the container and sits on the rim of the container. It allows users a simple and clean alternative to holding onto the specimen container itself. This handle is extremely beneficial for patients who may be pregnant or have limited mobility.