Water, Molecular Biology Grade, sterile RNase Free, DNase Free, Protease Free, DEPC Free

Supplier: Ricca Chemical

Synonyms: Dihydrogen oxide

R9145000-1G
75799-280CS 671.65 USD
75799-280
Water, Molecular Biology Grade, sterile RNase Free, DNase Free, Protease Free, DEPC Free
Water

Suitable for molecular and cell biology specifications.


  • RNase, DNase, Protease, Endotoxin-Free
  • DEPC Free
  • Sterile

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 0.997 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Appearance Clear, colorless liquid
Bioburden (no growth) To Pass Test
DNase Activity (none detected) To Pass Test
Endotoxin (none detected) To Pass Test
Protease Activity (none detected) To Pass Test
RNase Activity (none detected) To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR