Oil Return Kit, Labconco
Supplier: Labconco
1473500
75796-646EA 425 USD
75796-646
Oil Return Kit, Labconco
Tool Sets
Adjustable Gas Ballast Oil Return Kit for field installation on Labconco Rotary Vane Vacuum Pumps (sold separately).
It returns oil from exhaust filter housing to pump. This is needed when pump is run with the gas ballast open for extended periods of time.
Ordering information: Kit includes one oil return assembly, one drain adaptor, one restrictor, one flexible oil-return tube, two hose clips, one ‘O’ ring: Ø 14.6×2.4 mm, one ‘O’ ring: Ø 21.5×3.0 mm, and installation instructions.