Tuberculin Syringe, B. Braun
Supplier: B Braun
9161502SUS
75796-282CS 802.68 USD
75796-282
Tuberculin Syringe, B. Braun
Syringes Syringes, Medical
Syringe with consistent, dependable performance in a wide range of options.
  • Double seal stopper to prevent medication leakage
  • Large, stable finger grips
  • DEHP-free and latex-free
Smooth plunger action, high visibility - transparent barrel with bold graduations, secure backstop to prevent accidental plunger withdrawal.