Temperature Probe for Sonifier® SFX and 450D Series, Branson Ultrasonics
Supplier: Branson Ultrasonics
200-060-022R
75795-830EA 631.05 USD
75795-830
Temperature Probe for Sonifier® SFX and 450D Series, Branson Ultrasonics
Temperature Probes
The Temperature Probe accessory is a must-have for monitoring the temperature of sensitive samples
With Branson’s industry-exclusive True Temperature Control, the SFX Sonifiers® will automatically adjust the ultrasonic pulse length to maintain a temperature within a set range. Simply set a Pulse Temperature (the lower limit of the desired temperature range) and a Max. Temperature (upper limit), and the Sonifier® does the rest.
Simply plug the probe into the back of the Digital Sonifier®, and position its tip at the desired depth within a sample.