VAI® MP Garments, Veltek
Supplier: Veltek Associates
1700P-E-S-17000 1700P-E-S-17001 1700P-E-S-17002 1700P-E-S-17003 1700P-E-S-17004 1700P-E-S-17005 1700P-E-S-17006 1700P-H-UC-S-17017 1700P-H-UC-S-17035 1700-LC-S-1720 1700-LC-S-1725 1700P-E-S-17007 1700-LC-S-1724 1700-PS-S-1792 1700-LC-S-1721 1700-LC-S-1722 1700-LC-S-1723 1700P-HM-S-17037 1700P-HM-S-17038 1700P-MC-NS-S-17039 1700P-MC-NS-S-17040 1700P-MC-NS-S-17041
75779-672CS 415 USD
75779-672 75779-674 75779-676 75779-678 75779-680 75779-694 75779-696 75795-354 75795-402 75795-404 75795-408 75795-544 75795-584 75795-636 75795-642 75795-644 75795-646 76308-418 76308-420 76308-422 76308-424 76308-426
VAI® MP Garments, Veltek
Disposable Garments
The MP line is constructed using a non-woven, microporous, coated material that provides even greater personal protection in the cleanroom.
  • Highly breathable fabric releases heat while maintaining a high level of protection
  • Garments feature elastic in high movement areas for ease of movement and appropriate fit
  • 100% bound seam construction for increased durability and reduced particle permeability
  • Individually packaged in easy to tear bags with two liner bags per case
  • Packaging is labled with lot number and expiration date
All MP coveralls are equipped with elastic wrists and ankles, along with a zipper closure. All MP frocks feature elastic wrists and a zipper closure. The breathable fabric provides protection against low level splashes and sprays, offering reliable fluid and liquid resistance. All sterile garments are gamma irradiated at 10⁻⁶ SAL.
Ordering information: All garments are supplied with a one-year warranty.
Packaging: All garments are individually packaged, with the exception of the MP Knee-High boots, which are packaged in pairs.