Staticmaster® 1U400 Ionizing Cartridge, 1", NRD
Supplier: NRD Staticmaster
1U400
75794-964EA 289.87 USD
75794-964
Staticmaster® 1U400 Ionizing Cartridge, 1", NRD
Ionizers
The Staticmaster® 1U400 Ionizing Cartridge is a self-powered static eliminator with a small profile, making it an ideal solution for in-balance elimination
  • Self-powered
  • Small profile
  • Easy mounting
  • Ensures accurate weighing
  • 302 stainless steel housing and aluminum grid
This small profile ionizer offers "zero-volts balance" technology, which means that it can effectively remove all of the static charge present in your media and samples. This ensures accurate weighing every time.