Aluminized Apron, X20, Newtex Industries
Supplier: Newtex Industries
2100430
75794-934CS 2387.68 USD
75794-934
Aluminized Apron, X20, Newtex Industries
Workwear
Aluminized aprons for radiant heat protection.
THe X20 Apron features a Z-Flex A-601 aluminized outershell and a fire resistant cotton liner
Bib aprons are designed to provide protection in environments with ambient temperatures up to 200°F. Aprons are specifically engineered to maximize mobility and ease of use while offering the maximum protection and safeguards.
The outer shell reflects up to 95% of radiant heat while protecting from sparks, slag, and molten metal splash.