Staticmaster® Electrostatic Discharge Workstation Kit, NRD
Supplier: NRD Staticmaster
5021
75794-918EA 336.63 USD
75794-918
Staticmaster® Electrostatic Discharge Workstation Kit, NRD
Ionizers
The Staticmaster® 5021 ESD workstation kit provides an ESD safe work surface and grounding supplies for handling static sensitive products.
  • Provides ESD safe work surface
  • Includes grounded rubber table mat
The kit includes a 16" x 24" ESD rubber mat which has a dissipative top layer and a conductive bottom layer. The mat is grounded with a low profile common point ground and installed ground snap. A blue fabric wrist band and 6' coil cord grounds the operator.
Certifications: The kit meets or exceeds requirements of ANSI ESD S541-2003. RoHS Compliant.