Staticmaster® Electrostatic Discharge Workstation Kit with 1" Static Eliminating Brush, NRD
Supplier: NRD Staticmaster
SMM-KIT
75794-898KT 671.76 USD
75794-898
Staticmaster® Electrostatic Discharge Workstation Kit with 1" Static Eliminating Brush, NRD
Ionizers
This Staticmaster® microscopic kit is ideal for keeping slides clean and static free and is also ideal when working with tissue samples
  • Useful for microscopic work
  • Keeps slides clean
Includes (1) Staticmaster® 1C200 1" brush.