Anti-IgG Goat Antibody (AzureSpectra 550)

Supplier: Azure Biosystems
AC2160
75794-878EA 149.86 USD
75794-878
Anti-IgG Goat Antibody (AzureSpectra 550)
Antibodies
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: AzureSpectra 550
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR