Azure Fluorescent Blot Blocking Buffer
Supplier: Azure Biosystems
AC2190
75794-864EA 298.02 USD
75794-864
Azure Fluorescent Blot Blocking Buffer
Buffers Electrophoresis Buffers Blocking Buffers
Blocking buffer optimized for use with Azure’s fluorescent (and chemiluminescent) Western blotting systems.
Blocking Buffer for Fluorescent Western blots. 50x concentrated solution sufficient for 50-100 blots.