G418 disulfate 50 mg/mL in aqueous solution

Supplier: IBI Scientific
Danger

IB02060
75794-854EA 189.03 USD
75794-854
G418 disulfate 50 mg/mL in aqueous solution
G418 disulphate

G418 disulfate solution is routinely used as a selection antibiotic in cell culture gene selection applications


  • Concentration: 50mg/ml in pure water
  • Cell culture grade
  • Sterile filtered and verified for No Microbial growth
  • Packaged in a clear plastic bottle


G418 solution is an aminoglycoside antiotic isolated from Micromonospora rhodorangea and is closely related to the gentamicins, most notably gentamicin B1.

Formula: C₂₀H₄₄N₄O₁₈S₂
Storage Temperature: Freezer
MDL Number: MFCD00058314
CAS Number: 108321-42-2
EINECS: 213-791-2

Order Now

Specification Test Results

Appearance Clear, colorless liquid
Sterile filtration No microbial growth


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR